Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143757.1 | NP_001137229.1 | 11 | 1811 | ||
NM_030626.2 | NP_085129.1 | 11 | 1658 | |||
XM_006717983.2 | XP_006718046.1 | 11 | 1971 | |||
XM_011540208.1 | XP_011538510.1 | 11 | 1741 | |||
XM_011540210.2 | XP_011538512.1 | 10 | 1566 | |||
XM_017016694.1 | XP_016872183.1 | 10 | 1545 | |||
GenBank mRNA | AB051461.1 | - | 11 | 1568 | ||
AK091472.1 | - | 5 | 1744 | |||
AK093250.1 | - | 11 | 1424 | |||
AK097418.1 | - | 11 | 1769 |