Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303247.1 | NP_001290176.1 | 8, 9 | 960 | ||
NM_001303248.1 | NP_001290177.1 | 6, 7 | 575 | |||
NM_025234.2 | NP_079510.1 | 8, 9 | 854 | |||
XM_011522094.1 | XP_011520396.1 | 8, 9 | 834 | |||
XM_017022637.1 | XP_016878126.1 | 8, 9 | 750 | |||
GenBank mRNA | AF100786.1 | - | 8, 9 | 715 | ||
AF127799.1 | - | 7, 8 | 703 | |||
AF309553.1 | - | 7, 8 | 699 | |||
AK024754.1 | - | 8, 9 | 726 | |||
BC010080.2 | - | 8, 9 | 718 | 4329599 | ||
BU528743.1 | - | 6, 7 | 440 | |||
CR457333.1 | - | 6, 7 | 650 |