Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317950.1 | NP_001304879.1 | 21 | 4259 | ||
NM_001317952.1 | NP_001304881.1 | 20 | 3817 | |||
NM_030767.5 | NP_110394.3 | 21 | 4204 | |||
XM_005252244.2 | XP_005252301.1 | 23 | 4376 | |||
XM_005252245.1 | XP_005252302.1 | 22 | 4234 | |||
XM_005252247.4 | XP_005252304.1 | 21 | 4255 | |||
XM_005252248.2 | XP_005252305.1 | 20 | 4276 | |||
XM_006717294.1 | XP_006717357.1 | 21 | 4188 | |||
XM_011519063.2 | XP_011517365.1 | 20 | 3867 | |||
XM_011519065.2 | XP_011517367.2 | 21 | 3847 | |||
XM_017015172.1 | XP_016870661.1 | 22 | 3974 | |||
GenBank mRNA | AF286341.1 | - | 11 | 1758 | ||
AK024431.1 | - | 19 | 3152 | |||
AK074040.1 | - | 20 | 3247 | |||
AK131082.2 | - | 21 | 4260 | |||
AK160382.1 | - | 21 | 4262 | |||
AL713755.1 | - | 4 | 2693 | |||
AL833220.1 | - | 18 | 2681 | |||
AY703039.1 | - | 22 | 3859 | |||
AY703040.1 | - | 20 | 3817 | |||
AY703043.1 | - | 20 | 4177 | |||
AY703044.1 | - | 19 | 3871 | |||
AY703045.1 | - | 12 | 1950 | |||
AY703046.1 | - | 8 | 1076 | |||
BC042202.1 | - | 7 | 934 | |||
BC055285.1 | - | 21 | 4204 | 6143155 |