Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317950.1 | NP_001304879.1 | 18 | 3761 | ||
NM_001317952.1 | NP_001304881.1 | 17 | 3319 | |||
NM_030767.5 | NP_110394.3 | 18 | 3706 | |||
XM_005252244.2 | XP_005252301.1 | 20 | 3878 | |||
XM_005252245.1 | XP_005252302.1 | 19 | 3736 | |||
XM_005252247.4 | XP_005252304.1 | 18 | 3757 | |||
XM_005252248.2 | XP_005252305.1 | 18 | 3874 | |||
XM_006717294.1 | XP_006717357.1 | 18 | 3690 | |||
XM_011519063.2 | XP_011517365.1 | 17 | 3369 | |||
XM_011519064.2 | XP_011517366.1 | 18 | 3874 | |||
XM_011519065.2 | XP_011517367.2 | 18 | 3349 | |||
XM_017015172.1 | XP_016870661.1 | 19 | 3476 | |||
GenBank mRNA | AF286341.1 | - | 8 | 1261 | ||
AK024431.1 | - | 16 | 2654 | |||
AK074040.1 | - | 17 | 2749 | |||
AK131082.2 | - | 18 | 3761 | |||
AK160382.1 | - | 18 | 3763 | |||
AL713755.1 | - | 2 | 136 | |||
AL833220.1 | - | 15 | 2183 | |||
AY703039.1 | - | 19 | 3361 | |||
AY703040.1 | - | 17 | 3319 | |||
AY703043.1 | - | 17 | 3679 | |||
AY703044.1 | - | 16 | 3373 | |||
AY703045.1 | - | 9 | 1456 | |||
AY703046.1 | - | 5 | 582 | |||
BC042202.1 | - | 4 | 436 | |||
BC055285.1 | - | 18 | 3706 | 6143155 |