Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308207.1 | NP_001295136.1 | 5 | 1084 | ||
NM_001308208.1 | NP_001295137.1 | 4 | 737 | |||
NM_025251.2 | NP_079527.1 | 3 | 610 | |||
XM_011517308.1 | XP_011515610.1 | 4 | 891 | |||
XM_011517309.1 | XP_011515611.1 | 4 | 795 | |||
XM_011517312.2 | XP_011515614.1 | 2 | 1429 | |||
XM_017013870.1 | XP_016869359.1 | 5 | 1963 | |||
XM_017013871.1 | XP_016869360.1 | 4 | 1022 | |||
GenBank mRNA | AB051475.1 | - | 5 | 1088 | ||
AK296353.1 | - | 4 | 734 |