Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC44A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178045.1 | NP_001171516.1 | 6, 7 | 534 | ||
NM_025257.2 | NP_079533.2 | 6, 7 | 531 | |||
GenBank mRNA | AK027397.1 | - | 6, 7 | 482 | ||
AK222998.1 | - | 6, 7 | 482 | |||
AK223013.1 | - | 6, 7 | 482 | |||
AK300550.1 | - | 6, 7 | 534 | |||
AK314673.1 | - | 6, 7 | 486 | |||
AL833009.1 | - | 2, 3 | 224 | |||
BC014659.1 | - | 6, 7 | 470 | 4644272 | ||
DC300771.1 | - | 6, 7 | 531 | |||
KU175229.1 | - | 6, 7 | 465 |