Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174101.1 | NP_001167572.1 | 3 | 946 | ||
NM_001174102.1 | NP_001167573.1 | 3 | 863 | |||
NM_030567.4 | NP_085044.2 | 4 | 1031 | |||
XM_011534662.1 | XP_011532964.1 | 3 | 6769 | |||
XM_011534663.2 | XP_011532965.1 | 3 | 1507 | |||
XM_017009896.1 | XP_016865385.1 | 2 | 7557 | |||
XM_017009897.1 | XP_016865386.1 | 3 | 1739 | |||
GenBank mRNA | BC004261.1 | - | 4 | 863 | 3606982 | |
BC021240.1 | - | 3 | 854 | 3603954 | ||
BC024233.2 | - | 4 | 1035 | 4697841 | ||
BC033649.1 | - | 2 | 621 | 5538503 |