Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174101.1 | NP_001167572.1 | 3 | 1361 | ||
NM_001174102.1 | NP_001167573.1 | 3 | 1278 | |||
NM_030567.4 | NP_085044.2 | 4 | 1446 | |||
XM_011534662.1 | XP_011532964.1 | 3 | 7184 | |||
XM_011534663.2 | XP_011532965.1 | 3 | 1922 | |||
XM_017009896.1 | XP_016865385.1 | 2 | 7972 | |||
XM_017009897.1 | XP_016865386.1 | 3 | 2154 | |||
GenBank mRNA | BC004261.1 | - | 4 | 1278 | 3606982 | |
BC021240.1 | - | 3 | 1269 | 3603954 | ||
BC024233.2 | - | 4 | 1450 | 4697841 | ||
BC033649.1 | - | 2 | 1034 | 5538503 |