Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008695.1 | NP_001008695.1 | 3 | 205 | ||
NM_030573.2 | NP_085050.2 | 2 | 379 | |||
GenBank mRNA | AK024609.1 | - | 1 | 475 | ||
AK025906.1 | - | 1 | 243 | |||
AK315457.1 | - | 2 | 364 | |||
BC004346.1 | - | 2 | 350 | 3633743 | ||
BE264280.1 | - | 2 | 278 | |||
BI223500.1 | - | 2 | 322 | |||
BM790105.1 | - | 3 | 205 | |||
CR456343.1 | - | 2 | 197 |