Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000396.3 | NP_000387.1 | 4 | 606 | ||
GenBank mRNA | AK130134.1 | - | 4 | 425 | ||
AK308229.1 | - | 3 | 267 | |||
AK312880.1 | - | 4 | 423 | |||
AU118043.1 | - | 4 | 502 | |||
AU119985.1 | - | 4 | 501 | |||
BC016058.1 | - | 4 | 488 | 4860926 | ||
CR541675.1 | - | 3 | 377 | |||
CR541719.1 | - | 3 | 377 | |||
S79895.1 | - | 4 | 518 | |||
U13665.1 | - | 4 | 482 | |||
U20280.1 | - | 4 | 395 |