Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077484.1 | NP_001070952.1 | 11 | 1511 | ||
NM_001278387.1 | NP_001265316.1 | 12 | 1973 | |||
NM_001278388.1 | NP_001265317.1 | 10 | 1735 | |||
NM_001278389.1 | NP_001265318.1 | 11 | 1000 | |||
NM_001278390.1 | NP_001265319.1 | 11 | 1713 | |||
NM_030674.3 | NP_109599.3 | 11 | 1850 | |||
XM_011538784.2 | XP_011537086.1 | 11 | 1235 | |||
XM_011538786.2 | XP_011537088.1 | 11 | 1235 | |||
XM_011538787.2 | XP_011537089.1 | 11 | 1235 | |||
XM_017019990.1 | XP_016875479.1 | 11 | 1465 | |||
GenBank mRNA | AF247166.1 | - | 11 | 1196 | ||
AF271070.1 | - | 11 | 1094 | |||
AK027825.1 | - | 11 | 1000 | |||
AK074758.1 | - | 4 | 267 | |||
AK074797.1 | - | 8 | 569 | |||
AK074851.1 | - | 9 | 712 | |||
AK074949.1 | - | 9 | 758 | |||
AK225639.1 | - | 11 | 1088 | |||
AK225778.1 | - | 11 | 1110 | |||
AK308129.1 | - | 9 | 798 | |||
AK309410.1 | - | 11 | 1427 | |||
AK313731.1 | - | 9 | 801 | |||
BC010620.1 | - | 11 | 1057 | 3871101 | ||
BX647547.1 | - | 12 | 1545 | |||
BX648495.1 | - | 11 | 1438 | |||
BX648676.1 | - | 10 | 1305 |