Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025579.2 | NP_001020750.1 | 8 | 1048 | ||
NM_030808.4 | NP_110435.1 | 8 | 1048 | |||
XM_017025183.1 | XP_016880672.1 | 9 | 1228 | |||
XM_017025184.1 | XP_016880673.1 | 9 | 1228 | |||
XM_017025185.1 | XP_016880674.1 | 8 | 1097 | |||
XM_017025186.1 | XP_016880675.1 | 9 | 1228 | |||
XM_017025187.1 | XP_016880676.1 | 8 | 1097 | |||
GenBank mRNA | AF038203.1 | - | 2 | 147 | ||
AF182078.1 | - | 8 | 975 | |||
AF217798.3 | - | 8 | 1039 | |||
AK056014.1 | - | 8 | 1046 | |||
AK299559.1 | - | 9 | 1022 | |||
AL832648.1 | - | 8 | 982 | |||
AY004871.1 | - | 8 | 974 | |||
BC026101.2 | - | 8 | 986 | 4820855 | ||
DA256375.1 | - | 4 | 389 |