Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012514.2 | NP_001012532.1 | 4 | 597 | ||
NM_001012516.2 | NP_001012534.1 | 4 | 572 | |||
NM_001287240.1 | NP_001274169.1 | 5 | 674 | |||
NM_001287241.1 | NP_001274170.1 | 6 | 870 | |||
NM_030926.5 | NP_112188.1 | 5 | 738 | |||
GenBank mRNA | AB003629.1 | - | 5 | 607 | ||
AF271781.1 | - | 5 | 723 | |||
AF272043.1 | - | 5 | 720 | |||
AK056321.1 | - | 4 | 544 | |||
AK075361.1 | - | 5 | 703 | |||
AK090975.1 | - | 5 | 649 | |||
AK126739.1 | - | 1 | 1361 | |||
AK222697.1 | - | 5 | 685 | |||
AK295503.1 | - | 5 | 504 | |||
AK303199.1 | - | 5 | 692 | |||
AK307702.1 | - | 4 | 506 | |||
AL136603.1 | - | 5 | 679 | |||
AL713651.1 | - | 5 | 662 | |||
AY049777.1 | - | 5 | 624 | |||
BC002424.1 | - | 5 | 665 | 3162430 | ||
BC025742.1 | - | 4 | 542 | 5224043 | ||
BC050668.1 | - | 4 | 532 | 5759920 | ||
BC098563.1 | - | 5 | 651 | 30558635 | ||
BX444331.2 | - | 5 | 672 | |||
CR749389.1 | - | 1 | 1319 |