Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIGD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243253.1 | NP_001230182.1 | 2 | 1194 | ||
NM_030953.3 | NP_112215.1 | 2 | 1207 | |||
GenBank mRNA | AK056604.1 | - | 2 | 695 | ||
AK092978.1 | - | 2 | 1194 | |||
AK096325.1 | - | 2 | 1207 | |||
AL136539.1 | - | 2 | 618 | |||
BC033641.1 | - | 2 | 1154 | |||
BC117249.1 | - | 1 | 508 | 40125800 | ||
CR533559.1 | - | 1 | 432 |