Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324512.1 | NP_001311441.1 | 17 | 3111 | ||
NM_030955.3 | NP_112217.2 | 19 | 3366 | |||
XM_017009905.1 | XP_016865394.1 | 20 | 3465 | |||
XM_017009906.1 | XP_016865395.1 | 19 | 2866 | |||
XM_017009907.1 | XP_016865396.1 | 12 | 1758 | |||
XM_017009908.1 | XP_016865397.1 | 9 | 1350 | |||
XM_017009909.1 | XP_016865398.1 | 8 | 1264 | |||
GenBank mRNA | AJ250725.1 | - | 19 | 3353 | ||
BC131733.1 | - | 17 | 2860 | 40124828 |