Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127711.2 | NP_001121183.1 | 5 | 569 | ||
NM_001286484.1 | NP_001273413.1 | 5 | 594 | |||
NM_030969.4 | NP_112231.3 | 6 | 671 | |||
NR_104454.1 | 4 | 492 | ||||
GenBank mRNA | AK291883.1 | - | 6 | 625 | ||
BC001033.1 | - | 6 | 614 | 3139565 | ||
BC007080.1 | - | 6 | 626 | 4066217 | ||
BC013913.1 | - | 6 | 633 | 3685635 | ||
BC071660.1 | - | 6 | 615 | 4473732 | ||
BU954633.1 | - | 4 | 434 |