Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH11Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278619.1 | NP_001265548.1 | 6 | 3744 | ||
NM_032971.2 | NP_116753.1 | 5 | 3575 | |||
NM_032972.2 | NP_116754.1 | 2 | 3829 | |||
NM_032973.2 | NP_116755.1 | 2 | 3829 | |||
XM_017030079.1 | XP_016885568.1 | 8 | 8106 | |||
XM_017030080.1 | XP_016885569.1 | 7 | 4058 | |||
XM_017030081.1 | XP_016885570.1 | 6 | 3828 | |||
XM_017030082.1 | XP_016885571.1 | 6 | 3641 | |||
XM_017030083.1 | XP_016885572.1 | 3 | 3405 | |||
GenBank mRNA | AF277053.1 | - | 6 | 3675 | ||
AF332216.1 | - | 2 | 3829 | |||
AF332217.1 | - | 2 | 3829 | |||
AJ276803.1 | - | 5 | 3575 | |||
AJ564960.1 | - | 1 | 2459 | |||
AJ564961.1 | - | 1 | 2459 |