Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMFBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160213.1 | NP_001153685.1 | 7 | 1009 | ||
NM_031293.2 | NP_112583.2 | 6 | 831 | |||
XM_011523357.2 | XP_011521659.1 | 6 | 921 | |||
XM_011523358.2 | XP_011521660.1 | 6 | 861 | |||
XM_011523360.2 | XP_011521662.1 | 6 | 884 | |||
XM_011523361.2 | XP_011521663.1 | 6 | 1359 | |||
XM_011523362.2 | XP_011521664.1 | 5 | 666 | |||
GenBank mRNA | AK093285.1 | - | 6 | 827 | ||
AK097465.1 | - | 2 | 68 | |||
AK122925.1 | - | 7 | 1009 | |||
AK315004.1 | - | 6 | 812 | |||
AL136795.1 | - | 6 | 791 | |||
BC028423.2 | - | 6 | 818 | 4822328 |