Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142935.1 | NP_001136407.1 | 2, 3 | 642 | ||
NM_031300.3 | NP_112590.1 | 2, 3 | 642 | |||
GenBank mRNA | AF114834.1 | - | 2, 3 | 236 | ||
AK057034.1 | - | 2, 3 | 249 | |||
AK222579.1 | - | 2, 3 | 247 | |||
AK303397.1 | - | 2, 3 | 249 | |||
AK316468.1 | - | 2, 3 | 246 | |||
AL522759.3 | - | 2, 3 | 661 | |||
BC000745.2 | - | 2, 3 | 231 | 2821596 | ||
BC032586.1 | - | 2, 3 | 246 | |||
BM849150.1 | - | 2, 3 | 621 |