Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201366.1 | NP_001188295.1 | 7 | 548 | ||
NM_031427.3 | NP_113615.2 | 6 | 502 | |||
XM_017021679.1 | XP_016877168.1 | 7 | 536 | |||
GenBank mRNA | AF542071.1 | - | 6 | 378 | ||
AI242197.1 | - | 4 | 353 | |||
AK026653.1 | - | 3 | 160 | |||
AK315392.1 | - | 6 | 397 | |||
AL833654.1 | - | 5 | 258 | |||
BC005343.1 | - | 6 | 378 | 3930213 | ||
BJ996719.1 | - | 2 | 98 |