Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTBDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080997.2 | NP_001074466.1 | 6 | 693 | ||
NM_001270440.1 | NP_001257369.1 | 7 | 1699 | |||
NM_001270441.1 | NP_001257370.1 | 6 | 646 | |||
NM_001270442.1 | NP_001257371.1 | 6 | 634 | |||
NM_001270443.1 | NP_001257372.1 | 6 | 622 | |||
NM_001270444.1 | NP_001257373.1 | 6 | 610 | |||
NM_001270445.1 | NP_001257374.1 | 6 | 604 | |||
NM_031429.2 | NP_113617.1 | 7 | 966 | |||
XM_005260089.1 | XP_005260146.1 | 7 | 1041 | |||
XM_005260090.3 | XP_005260147.1 | 6 | 904 | |||
XM_011528343.1 | XP_011526645.1 | 6 | 707 | |||
XM_011528344.2 | XP_011526646.1 | 6 | 757 | |||
XM_011528345.1 | XP_011526647.1 | 6 | 702 | |||
XM_017027343.1 | XP_016882832.1 | 7 | 996 | |||
XM_017027344.1 | XP_016882833.1 | 7 | 948 | |||
GenBank mRNA | AB593122.1 | - | 6 | 646 | ||
AK093672.1 | - | 7 | 1699 | |||
AY028917.1 | - | 6 | 670 | |||
BC005063.1 | - | 7 | 954 | 3344818 | ||
BG471112.1 | - | 6 | 607 | |||
BM688127.1 | - | 6 | 601 | |||
BQ639040.1 | - | 7 | 568 |