Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031431.3 | NP_113619.2 | 13 | 1615 | ||
XM_011535266.1 | XP_011533568.1 | 13 | 1422 | |||
XM_011535267.2 | XP_011533569.1 | 13 | 1595 | |||
XM_017020790.1 | XP_016876279.1 | 12 | 1268 | |||
GenBank mRNA | AF332595.1 | - | 13 | 1574 | ||
AF349676.1 | - | 13 | 1594 | |||
AK026305.1 | - | 13 | 1574 | |||
AK074374.1 | - | 1 | 159 | |||
AK225466.1 | - | 13 | 1574 | |||
AK225952.1 | - | 13 | 1574 | |||
AK304248.1 | - | 13 | 1417 | |||
AK314387.1 | - | 13 | 1541 | |||
BX648211.1 | - | 13 | 1178 |