Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000499.4 | NP_000490.1 | 7 | 1572 | ||
NM_001319216.1 | NP_001306145.1 | 6 | 1488 | |||
NM_001319217.1 | NP_001306146.1 | 7 | 1575 | |||
XM_017021953.1 | XP_016877442.1 | 8 | 1491 | |||
GenBank mRNA | AK223108.1 | - | 7 | 1493 | ||
AK223113.1 | - | 7 | 1490 | |||
AK310810.1 | - | 7 | 1619 | |||
AK313880.1 | - | 7 | 1490 | |||
AM236046.1 | - | 8 | 1376 | |||
AM236047.1 | - | 7 | 671 | |||
AY310359.1 | - | 7 | 1292 | |||
AY871801.1 | - | 6 | 1372 | |||
BC023019.1 | - | 7 | 1457 | 5123393 |