Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031442.3 | NP_113630.1 | 2, 3 | 630 | ||
GenBank mRNA | AK311054.1 | - | 2, 3 | 313 | ||
AL136550.1 | - | 2, 3 | 380 | |||
BC039242.1 | - | 2, 3 | 630 | 5277761 | ||
CX788638.1 | - | 2, 3 | 618 |