Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMMECR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199140.1 | NP_001186069.1 | 7 | 1056 | ||
NM_031445.2 | NP_113633.2 | 7 | 1028 | |||
XM_005263804.4 | XP_005263861.1 | 8 | 1659 | |||
XM_005263806.4 | XP_005263863.1 | 7 | 1548 | |||
XM_005263807.4 | XP_005263864.1 | 6 | 1413 | |||
XM_011511952.2 | XP_011510254.1 | 7 | 1549 | |||
XM_011511954.2 | XP_011510256.1 | 8 | 1636 | |||
XM_011511955.2 | XP_011510257.1 | 6 | 578 | |||
XM_011511956.2 | XP_011510258.1 | 5 | 1318 | |||
GenBank mRNA | AK095871.1 | - | 2 | 113 | ||
AK304145.1 | - | 7 | 1056 | |||
BC016181.1 | - | 7 | 1028 | 4644159 | ||
BC078174.1 | - | 7 | 1038 | 6012672 |