Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18orf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201474.1 | NP_001188403.1 | 4 | 629 | ||
NM_001201475.1 | NP_001188404.1 | 4 | 696 | |||
NM_001201476.1 | NP_001188405.1 | 4 | 426 | |||
NM_031446.4 | NP_113634.3 | 5 | 617 | |||
XM_005258364.4 | XP_005258421.1 | 5 | 622 | |||
GenBank mRNA | AF277182.1 | - | 5 | 476 | ||
AF277189.1 | - | 5 | 476 | |||
AY631401.1 | - | 3 | 249 | |||
BC025950.1 | - | 4 | 536 | |||
BC108730.1 | - | 5 | 577 | 5187778 | ||
BC119765.1 | - | 5 | 568 | 40119355 | ||
BC119766.1 | - | 5 | 567 | 40119357 | ||
BX460890.2 | - | 5 | 943 | |||
DN991579.1 | - | 4 | 349 |