Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIOK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031480.2 | NP_113668.2 | 10 | 1100 | ||
NM_153005.1 | NP_694550.1 | 3 | 410 | |||
XM_005249446.3 | XP_005249503.1 | 10 | 1145 | |||
XM_011514933.2 | XP_011513235.2 | 10 | 1062 | |||
GenBank mRNA | AF258661.1 | - | 3 | 410 | ||
AK054568.1 | - | 10 | 1091 | |||
AK314467.1 | - | 10 | 1129 | |||
AL834277.1 | - | 9 | 799 | |||
BC006104.1 | - | 10 | 1100 | 3537086 |