Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135105.1 | NP_001128577.1 | 4 | 631 | ||
NM_001135106.1 | NP_001128578.1 | 4 | 631 | |||
NM_001135107.1 | NP_001128579.1 | 4 | 631 | |||
NM_032115.3 | NP_115491.1 | 4 | 631 | |||
XM_011514935.2 | XP_011513237.1 | 5 | 687 | |||
XM_017011346.1 | XP_016866835.1 | 4 | 725 | |||
GenBank mRNA | AF358909.1 | - | 4 | 617 | ||
AY253145.1 | - | 4 | 617 | |||
AY253147.1 | - | 4 | 617 | |||
BC111860.1 | - | 4 | 617 | 40016308 | ||
EU978943.1 | - | 4 | 665 |