Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201466.1 | NP_001188395.1 | 3 | 417 | ||
NM_031916.4 | NP_114122.2 | 2 | 540 | |||
XM_006714503.2 | XP_006714566.1 | 2 | 514 | |||
XM_006714504.2 | XP_006714567.1 | 3 | 416 | |||
XM_017009946.1 | XP_016865435.1 | 3 | 416 | |||
XM_017009947.1 | XP_016865436.1 | 3 | 416 | |||
GenBank mRNA | AF239723.1 | - | 2 | 274 | ||
AK098642.1 | - | 2 | 429 | |||
BC014607.2 | - | 2 | 501 | 4820798 | ||
BX113042.1 | - | 3 | 382 | |||
CV024170.1 | - | 2 | 234 | |||
DB479784.1 | - | 3 | 417 |