Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP3-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031959.2 | NP_114165.1 | 1 | 565 | ||
GenBank mRNA | AJ406932.1 | - | 1 | 566 | ||
BC034582.1 | - | 1 | 573 | 3909612 |