Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP9-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031961.2 | NP_114167.2 | 1 | 851 | ||
GenBank mRNA | AJ406946.1 | - | 1 | 851 |
Gene Symbol: KRTAP9-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031963.2 | NP_114169.2 | 1 | 833 |