Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTTNBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033427.2 | NP_219499.1 | 22 | 4796 | ||
XM_005250634.4 | XP_005250691.1 | 22 | 4931 | |||
XM_005250635.3 | XP_005250692.1 | 22 | 4565 | |||
XM_011516615.2 | XP_011514917.1 | 22 | 7099 | |||
XM_017012706.1 | XP_016868195.1 | 22 | 4728 | |||
XM_017012708.1 | XP_016868197.1 | 19 | 4310 | |||
GenBank mRNA | AB051545.1 | - | 22 | 4702 | ||
AF377960.1 | - | 22 | 4796 | |||
AK096565.1 | - | 10 | 1333 | |||
BC106000.1 | - | 22 | 4763 | 30528261 |