Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077263.2 | NP_001070731.1 | 9 | 1360 | ||
NM_001206789.1 | NP_001193718.1 | 8 | 1255 | |||
NM_001206790.1 | NP_001193719.1 | 9 | 1360 | |||
NM_001244995.1 | NP_001231924.1 | 9 | 1360 | |||
GenBank mRNA | AB048796.1 | - | 9 | 1286 | ||
AB048797.1 | - | 8 | 1211 | |||
AK027798.1 | - | 9 | 1304 | |||
AK300283.1 | - | 9 | 1272 | |||
AY190317.1 | - | 9 | 1287 | |||
BC018715.1 | - | 10 | 1095 | |||
BC114928.1 | - | 9 | 1279 | 40028749 |