Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160184.1 | NP_001153656.1 | 15 | 2137 | ||
NM_032129.2 | NP_115505.2 | 16 | 2242 | |||
GenBank mRNA | AL136730.1 | - | 16 | 2242 | ||
BC101386.2 | - | 15 | 2132 | 40026400 | ||
BC101387.2 | - | 15 | 2393 | 40026404 |