Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QRICH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032134.2 | NP_115510.1 | 4 | 1050 | ||
XM_005257728.3 | XP_005257785.1 | 4 | 2060 | |||
XM_011525344.2 | XP_011523646.1 | 2 | 733 | |||
XM_017025206.1 | XP_016880695.1 | 4 | 2060 | |||
XM_017025207.1 | XP_016880696.1 | 3 | 2000 | |||
GenBank mRNA | AL136774.1 | - | 4 | 1050 |