Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032145.4 | NP_115521.3 | 2 | 745 | ||
XM_005267159.3 | XP_005267216.1 | 2 | 584 | |||
XM_011536177.2 | XP_011534479.1 | 2 | 5604 | |||
XM_017011353.1 | XP_016866842.1 | 2 | 584 | |||
GenBank mRNA | BC024326.1 | - | 2 | 713 | 3915249 |