Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032169.4 | NP_115545.3 | 8, 9 | 1442 | ||
NR_132426.1 | 8, 9 | 1442 | ||||
NR_132427.1 | 8, 9 | 1442 | ||||
NR_132428.1 | 8, 9 | 1442 | ||||
GenBank mRNA | AJ608287.1 | - | 8, 9 | 1393 | ||
AK022654.1 | - | 1, 2 | 36 | |||
AK131265.1 | - | 10, 11 | 1239 | |||
AK308380.1 | - | 9, 10 | 1032 | |||
AL832873.1 | - | 8, 9 | 1393 | |||
AL833721.1 | - | 8, 9 | 960 | |||
AX803783.1 | - | 8, 9 | 1299 | |||
AX803785.1 | - | 8, 9 | 1062 | |||
BC019607.1 | - | 3, 4 | 338 | |||
BC125204.1 | - | 8, 9 | 1377 | 40123454 | ||
BC125205.1 | - | 8, 9 | 1377 |
Gene Symbol: NPHP3-ACAD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037804.1 | 33, 34 | 4903 | |||
GenBank mRNA | AB082531.1 | - | 33, 34 | 4903 |