Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032188.2 | NP_115564.2 | 6 | 736 | ||
NM_182958.2 | NP_892003.2 | 6 | 736 | |||
XM_011545970.1 | XP_011544272.1 | 7 | 1277 | |||
XM_011545971.1 | XP_011544273.1 | 7 | 1282 | |||
GenBank mRNA | AF217501.1 | - | 6 | 702 | ||
AF260665.2 | - | 6 | 634 | |||
AF289578.1 | - | 3 | 1664 | |||
AK021872.1 | - | 6 | 733 | |||
AK024102.1 | - | 6 | 734 | |||
AK225660.1 | - | 6 | 736 | |||
AK291106.1 | - | 6 | 733 | |||
AL050395.1 | - | 2 | 120 | |||
BC037773.1 | - | 6 | 733 | 5278026 |