Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ASCC2 SIRNA Assays ›
Gene Symbol:
ASCC2
Gene Name:
activating signal cointegrator 1 complex subunit 2
Gene Aliases:
ASC1p100, p100
Chromosome Location:
Chr.22: 29788608 - 29838304 on Build GRCh38
UniGene ID:
Hs.731754
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s38588
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ASCC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242906.1
NP_001229835.1
12
1177
NM_032204.4
NP_115580.2
13
1405
XM_005261775.2
XP_005261832.1
14
1445
XM_011530442.2
XP_011528744.1
14
1445
XM_011530443.2
XP_011528745.1
15
1511
XM_011530444.2
XP_011528746.1
15
1558
XM_011530445.2
XP_011528747.1
14
1492
XM_011530446.2
XP_011528748.1
13
1375
XM_011530448.2
XP_011528750.1
13
1278
XM_011530449.2
XP_011528751.1
12
1216
XM_011530450.2
XP_011528752.1
15
1561
XM_011530451.2
XP_011528753.1
16
1635
XM_011530452.2
XP_011528754.1
15
1608
XM_011530453.2
XP_011528755.1
14
1463
XM_011530454.2
XP_011528756.1
13
1347
XM_011530455.2
XP_011528757.1
12
1276
XM_017028991.1
XP_016884480.1
13
3583
XM_017028992.1
XP_016884481.1
13
3583
XM_017028993.1
XP_016884482.1
12
3508
XM_017028994.1
XP_016884483.1
12
3508
XM_017028995.1
XP_016884484.1
12
3424
XM_017028996.1
XP_016884485.1
14
2385
XM_017028997.1
XP_016884486.1
12
3424
XM_017028998.1
XP_016884487.1
14
2385
XM_017028999.1
XP_016884488.1
15
1511
XM_017029000.1
XP_016884489.1
14
1492
XM_017029001.1
XP_016884490.1
13
1278
XM_017029002.1
XP_016884491.1
12
1216
XM_017029003.1
XP_016884492.1
14
1352
XM_017029004.1
XP_016884493.1
11
1141
XM_017029005.1
XP_016884494.1
15
1561
XM_017029006.1
XP_016884495.1
16
1635
XM_017029007.1
XP_016884496.1
14
1463
XM_017029008.1
XP_016884497.1
13
1347
XM_017029009.1
XP_016884498.1
12
1276
GenBank mRNA
AK022886.1
-
15
1523
AK025241.1
-
5
434
AK222723.1
-
5
434
AK223033.1
-
5
433
AK303257.1
-
12
1147
AL096741.1
-
1
95
AY013289.1
-
12
1282
BC025368.2
-
13
1366
3955747
Back To Top
More Information
Gene Ontology Categories:
Function(s)
DNA dealkylation involved in DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0