Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131035.1 | NP_001124507.1 | 6 | 907 | ||
NM_001131036.1 | NP_001124508.1 | 6 | 879 | |||
NM_032280.2 | NP_115656.1 | 6 | 1264 | |||
XM_005248616.3 | XP_005248673.1 | 5 | 477 | |||
GenBank mRNA | AK314160.1 | - | 6 | 1207 | ||
AL512712.1 | - | 5 | 3928 | |||
BC032736.1 | - | 6 | 844 | 5540043 |