Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC37A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287498.1 | NP_001274427.1 | 3 | 312 | ||
NM_032295.3 | NP_115671.1 | 3 | 312 | |||
NM_207113.2 | NP_996996.1 | 3 | 312 | |||
XM_011516626.2 | XP_011514928.1 | 5 | 809 | |||
XM_011516627.2 | XP_011514929.1 | 5 | 808 | |||
XM_017012712.1 | XP_016868201.1 | 5 | 785 | |||
XM_017012713.1 | XP_016868202.1 | 5 | 804 | |||
XM_017012714.1 | XP_016868203.1 | 5 | 825 | |||
XM_017012715.1 | XP_016868204.1 | 5 | 828 | |||
XM_017012716.1 | XP_016868205.1 | 5 | 832 | |||
XM_017012717.1 | XP_016868206.1 | 5 | 804 | |||
XM_017012718.1 | XP_016868207.1 | 5 | 803 | |||
GenBank mRNA | AK074823.1 | - | 3 | 273 | ||
AK296543.1 | - | 3 | 266 | |||
AK299276.1 | - | 3 | 256 | |||
AK300265.1 | - | 3 | 270 | |||
AL136583.1 | - | 3 | 219 | |||
BC028380.1 | - | 3 | 262 | 4811814 | ||
BC046567.1 | - | 3 | 264 | 5246993 | ||
CD101739.1 | - | 3 | 320 |