Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCHP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143852.1 | NP_001137324.1 | 11 | 1350 | ||
NM_032300.4 | NP_115676.1 | 11 | 1411 | |||
XM_011538836.2 | XP_011537138.1 | 11 | 1686 | |||
XM_011538837.2 | XP_011537139.1 | 11 | 1259 | |||
XM_017020024.1 | XP_016875513.1 | 10 | 1268 | |||
XM_017020025.1 | XP_016875514.1 | 11 | 1595 | |||
GenBank mRNA | AK092736.1 | - | 11 | 1350 | ||
AK301262.1 | - | 7 | 784 | |||
AY007230.1 | - | 11 | 1412 | |||
BC004285.1 | - | 11 | 1331 | 3617849 | ||
BX648596.1 | - | 1 | 299 |