Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134395.1 | NP_001127867.1 | 5 | 1198 | ||
NM_001134396.1 | NP_001127868.1 | 5 | 1179 | |||
NM_001318252.1 | NP_001305181.1 | 5 | 2082 | |||
NM_032350.5 | NP_115726.1 | 5 | 1208 | |||
NR_134537.1 | 4 | 1184 | ||||
XM_006715793.2 | XP_006715856.1 | 5 | 1610 | |||
XM_017012722.1 | XP_016868211.1 | 6 | 1029 | |||
GenBank mRNA | AF495721.1 | - | 5 | 2038 | ||
BC006224.2 | - | 5 | 1208 | 3941780 |