Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032353.3 | NP_115729.1 | 3 | 266 | ||
XM_017025224.1 | XP_016880713.1 | 3 | 232 | |||
GenBank mRNA | AB014763.1 | - | 3 | 226 | ||
AK312092.1 | - | 3 | 253 | |||
BC006282.2 | - | 3 | 266 | 4025884 |