Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021080.3 | NP_066566.3 | 8 | 1138 | ||
GenBank mRNA | AB210012.1 | - | 8 | 1120 | ||
AK095513.1 | - | 8 | 1138 | |||
AK313144.1 | - | 6 | 785 | |||
BC067445.1 | - | 5 | 500 | 6971707 | ||
BC067446.1 | - | 5 | 500 | 6971709 | ||
BC112938.1 | - | 6 | 1107 | 4510753 |