Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032427.3 | NP_115803.1 | 5 | 4495 | ||
XM_011543023.2 | XP_011541325.1 | 5 | 6330 | |||
XM_011543024.2 | XP_011541326.1 | 5 | 3035 | |||
GenBank mRNA | AB058722.1 | - | 5 | 4486 | ||
AY040322.1 | - | 5 | 4486 | |||
BC143529.1 | - | 5 | 3236 | 9052038 | ||
BC152449.1 | - | 5 | 4486 | 8862580 | ||
CR627398.1 | - | 1 | 3491 |