Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252675.1 | NP_001239604.1 | 13 | 1879 | ||
NM_001252676.1 | NP_001239605.1 | 12 | 1712 | |||
NM_032501.3 | NP_115890.2 | 13 | 1885 | |||
XM_006723659.1 | XP_006723722.1 | 9 | 1231 | |||
XM_011529388.1 | XP_011527690.1 | 12 | 1670 | |||
GenBank mRNA | AB058749.1 | - | 8 | 802 | ||
AK027817.1 | - | 7 | 708 | |||
AK090851.1 | - | 1 | 645 | |||
AK092295.1 | - | 11 | 1294 | |||
AK125058.1 | - | 11 | 3088 | |||
AK125923.1 | - | 1 | 2972 | |||
AK296306.1 | - | 12 | 1709 | |||
AK314440.1 | - | 13 | 1808 | |||
AL832939.1 | - | 9 | 954 | |||
AL833041.1 | - | 1 | 138 | |||
BC032076.1 | - | 6 | 573 | |||
BC039261.1 | - | 13 | 1844 | 5298849 | ||
BC044588.1 | - | 13 | 1804 | 5260496 | ||
BC055008.1 | - | 13 | 1804 | 4838976 |