Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142351.1 | NP_001135823.1 | 2 | 441 | ||
NM_001142352.1 | NP_001135824.1 | 2 | 511 | |||
NM_001322362.1 | NP_001309291.1 | 2 | 490 | |||
NM_032528.2 | NP_115917.1 | 2 | 511 | |||
XM_006712802.1 | XP_006712865.1 | 2 | 526 | |||
XM_011511999.2 | XP_011510301.1 | 2 | 623 | |||
XM_011512000.2 | XP_011510302.1 | 2 | 708 | |||
XM_011512001.2 | XP_011510303.1 | 2 | 708 | |||
XM_017005110.1 | XP_016860599.1 | 2 | 462 | |||
GenBank mRNA | AB058780.1 | - | 2 | 441 | ||
AB059555.1 | - | 2 | 497 | |||
AK307784.1 | - | 2 | 512 | |||
BC008680.1 | - | 2 | 497 | 3856171 |