Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308096.1 | NP_001295025.1 | 14 | 1434 | ||
NM_001324096.1 | NP_001311025.1 | 13 | 1277 | |||
NM_001324097.1 | NP_001311026.1 | 13 | 1295 | |||
NM_001324098.1 | NP_001311027.1 | 13 | 1487 | |||
NM_032569.3 | NP_115958.2 | 14 | 1452 | |||
NR_136695.1 | 15 | 1737 | ||||
NR_136696.1 | 14 | 1520 | ||||
NR_136697.1 | 14 | 1538 | ||||
NR_136698.1 | 13 | 1469 | ||||
NR_136699.1 | 13 | 1487 | ||||
NR_136700.1 | 13 | 1604 | ||||
XM_005255639.4 | XP_005255696.2 | 12 | 1202 | |||
XM_011522716.2 | XP_011521018.1 | 14 | 1580 | |||
XM_011522717.1 | XP_011521019.1 | 15 | 1637 | |||
XM_017023786.1 | XP_016879275.1 | 14 | 1530 | |||
XM_017023787.1 | XP_016879276.1 | 13 | 1464 | |||
XM_017023788.1 | XP_016879277.1 | 15 | 1619 | |||
XM_017023789.1 | XP_016879278.1 | 14 | 1586 | |||
GenBank mRNA | AF244907.1 | - | 14 | 1433 | ||
AF326966.1 | - | 14 | 1397 | |||
AK296842.1 | - | 13 | 1186 | |||
AK307834.1 | - | 14 | 1427 | |||
AY352585.1 | - | 14 | 1415 | |||
BC003693.1 | - | 12 | 1326 | |||
BC032855.1 | - | 14 | 1443 | 5265561 | ||
BC047223.1 | - | 14 | 1432 | 5263991 | ||
BC064940.1 | - | 15 | 1636 | 5787345 |