Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026761.2 | 3, 4 | 395 | |||
GenBank mRNA | AK130857.1 | - | 3, 4 | 362 | ||
AK130858.1 | - | 3, 4 | 347 | |||
BC005997.1 | - | 3, 4 | 331 | |||
BP197632.1 | - | 3, 4 | 396 | |||
BX382054.2 | - | 3, 4 | 330 |